Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSPT2 Mouse, Unlabeled, Clone: 32, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611250

Catalog No. BDB611250


Add to cart

Description

Description

Cell cycle progression is regulated by multiple genes at the G1 to S phase transition. The yeast GST1 gene encodes a GTP-binding protein that can rescue the G1 to S phase transition. This function is conserved in the GST1 human homolog, GSPT1. GSPT1 is also thought to function in protein synthesis as a polypeptide chain releasing factor, eRF3. eRF3 facilitates ribosomal binding of eRF1, a factor which induces ribosomal release of the nascent peptide. In addition to a GSPT1 homolog, a second GSPT protein (GSPT2) has been identified in
mice. These proteins have a unique N-terminal region, four conserved GTP- binding domains, and a C-terminal domain with homology to elongation factor-1α(EF1α). Although both GSPTs are expressed in a wide range of tissues, GSPT2 levels are elevated in the brain. GSPT1 expression transiently increases during the G1 to S phase transition, while GSPT2 levels remain constant during all phases of the cell cycle. Both proteins can bind eRF1 and may therefore function as eRF3. Thus, the GSPT proteins play important roles in cell cycle regulation and translation termination and may serve to link these essential biological processes.

Host Species: Mouse
Clone: 32
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Mouse GSPT2 aa. 19-126
Formula Weight [Chemical]: 88/90kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

GSPT2
32
Unconjugated
Murine
IgG1
RUO
Primary
Murine, Rat
Immunofluorescence, Western Blot
250μg/mL
Affinity Purified
Mouse GSPT2 aa. 19-126
50μg
Cell Biology
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit