Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-GST Epitope Tag, Agarose, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP136792

Catalog No. NBP136792

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

GST Epitope Tag
0.25 mg/mL
Immunoprecipitation 1:10-1:500
Affinity Purified
GSTM1
Rabbits were immunized with Glutathione-S-Transferase (GST) from Schistosoma japonicum. Antibody was isolated by affinity chromatography using GST immobilized on solid support.
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Non-species specific
Immunoprecipitation
Agarose
PBS with 0.09% Sodium Azide
EC 2.5.1.18, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, glutathione S-aryltransferase, glutathione S-transferase M1, glutathione S-transferase mu 1, GST class-mu 1, GST HB subunit 4, GST1, GSTM1-1, GSTM1a-1a, GSTM1b-1b, GTH4, GTM1, H-B, HB subunit 4, MGC26563, MU, MU-1, S-(hydroxyalkyl)glutathione lyase
Rabbit
IgG
0.1 mg
Epitope Tags, GST Epitope Tags
Primary
2944
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only