Learn More
anti-GST HRP Conjugate, GE Healthcare: PROMO
This highly specific antibody to GST is conjugated to horseradish peroxidase and optimized for use in Western blotting with ECL Detection Reagents.
Manufacturer:  GE Healthcare Life Sciences RPN1236PM
Catalog No. 45-001-278LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Binds with high affinity to GST fusion proteins.
  • Detection of GST is simplified because no secondary antibody is required
  • ECL optimized protocols included.
  • The conjugate recognizes multiple epitopes of GST and therefore is not reliant on functional GST for fusion protein detection. It has also been cross-absorbed to E. coli proteins so that GST-fusion proteins can be easily and specifically identified against an E. coli background.
  • Offers the advantages of speed, sensitivity, increased safety, and convenience when used with ECL detection systems. Application tested with ECL and ECL Plus reagents in western blotting for sensitive hard copy results
  • Also suitable for colorimetric detection with diaminobenzidine (DAB) and tetramethylbenzidine (TMB).
GST is a commonly used expression marker in cloning that is also used for purification and detection of fusion proteins.
Specifications
Specifications
GST
Purified
Primary
HRP
75μL
Monoclonal