Learn More
UbpbioSupplier Diversity Partner ANTI-GST, MOUSE 100UG DFS Item
Manufacturer:  UbpbioSupplier Diversity Partner Y1021
Antibodies – Anti-GST (UBPBio catalog# Y1021) – Mouse monoclonal antibody (clone GST.B6, IgG2a). Used for immunoblotting at 1:1000-3000 dilution for detection of 10-50 ng GST-tagged proteins; immunostaining at 1:500-2,000 dilution. May also be used for dot, ELISA and IP assays.
Catalog No. 50-112-6917