Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GST Tag Mouse anti-All, Clone: DG122-2A7, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05311
Description
Detect GST Tag using this Anti-GST Tag Antibody, clone DG122-2A7 validated for use in immunoprecipitation and western blotting.Specifications
GST Tag | |
Monoclonal | |
Unconjugated | |
NM_000852.2 | |
Mouse | |
Protein G purfied | |
Primary | |
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles. | |
IgG |
Immunoprecipitation, Western Blot | |
DG122-2A7 | |
Protein G Purified immunoglobulin in Protein G Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105MNaCl, 0.035% sodium azide as a preservative. | |
GSTP1, DFN7, GST3, FAEES3, GSTP1-1, PI | |
GST dervied from the pGEX expression vector, linked to KLH. | |
250 μg | |
All species | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only