Learn More
GST Tag Mouse anti-Bacteria, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05782
Catalog No. 05-782-MI
Add to cart
Description
Description
Detect GST Tag using this Anti-GST Tag Antibody validated for use in western blotting.
Specifications
Specifications
GST Tag
Unconjugated
NM_000852.2
GSTP1, DFN7, GST3, FAEES3, GSTP1-1, PI
Recombinant GST (glutathione S-transferase) from Shistosoma japonicum expressed in E. coli
Protein G purfied
−20°C for 24 months.
Monoclonal
Western Blot
protein G purified mouse IgG in storage buffer (PBS containing 0.05% sodium azide) and 30% glycerol
Purified
Murine
IgG
200μg
Primary
Bacteria