Learn More
GST Tag Rabbit anti-All, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06332
Catalog No. 06-332-MI
Add to cart
Description
Description
Anti-GST Tag Antibody detects level of GST Tag and has been published and validated for use in western blotting.
Specifications
Specifications
GST Tag
Unconjugated
Purified
Rabbit
IgG
200μg
Primary
NM_000852.2
Western Blot
Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide as a preservative.
GSTP1; DFN7; GST3; FAEES3; GSTP1-1; PI
GST (Glutathione-S-transferase) derived from the pGEX expression vector and linked to KLH
Protein A purfied
−20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Polyclonal
All