Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GST Omega 1 140A, Polyclonal, Invitrogen™

Catalog No. PIPA127745
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA127745 50 μg
1 options

Catalog No. PIPA127745

Supplier: Thermo Scientific PA127745

Chicken Polyclonal Antibody

Exhibits glutathione-dependent thiol transferase and dehydroascorbate reductase activities. This protein is a member of the theta class glutathione S-transferase-like (GSTTL) protein family. In mouse. the encoded protein acts as a small stress response pr

PA1-27745 detects GSTO1 from human samples.PA1-27745 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27761 immunogen is a synthetic peptide corresponding to human Glutathione transferase omega-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTO1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene GSTO1
Gene Accession No. P78417
Gene Alias RP11-99N20.1, DKFZp686H13163, GSTTLp28, P28
Gene Symbols GSTO1
Host Species Chicken
Immunogen Synthetic peptide corresponding to human Glutathione transferase omega-1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9446
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.