Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTF2IRD2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524871

Catalog No. PIPA524871


Add to cart

Description

Description

GTD2A is unusual in that its coding sequence is mostly derived from Charlie8 repeat elements. However, there is mRNA and EST evidence to suggest that this gene is transcribed, and the encoded protein has a homolog in mouse, with which it shares 78% sequence identity. The exact function of this gene product is not known. It is inferred to be a transcription factor based on the presence of GTF2I-like repeats (containing helix-loop-helix motifs), also found in other proteins such as GTF2IRD1 and GTF2I. These three genes are clustered together on chr 7q11.23.
Specifications

Specifications

GTF2IRD2
Polyclonal
PBS with 0.09% sodium azide
Q86UP8
FP630, GTF2IRD2A, GTF2I repeat domain-containing protein 2A, GTF2IRD2 alpha, general transcription factor II i repeat domain 2 alpha, general transcription factor II-I repeat domain-containing protein 2A, transcription factor GTF2IRD2, transcription factor GTF2IRD2-alpha
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84163
Flow Cytometry, Western Blot
Unconjugated
GTF2IRD2
Liquid
GTF2IRD2
KLH conjugated synthetic peptide between 812-841 amino acids from the C-terminal region of human GTD2A
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit