Learn More
GTF2A1 Mouse anti-Human, Clone: 2H5, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA519032
Catalog No. PIMA519032
Add to cart
Description
Description
Peptide Sequence: MANSANTNTV PKLYRSVIED VINDVRDIFL DDGVDEQVLM ELKTLWENKL MQSRAVDGFH SEE
Transcription initiation factor IIA alpha; and beta; chains (GTF2A1, TFIIA p35 and p19 subunits, TFIIAL) binding ability of TLP is required for characteristic cytoplasmic localization of TLP. TFIIA may regulate the intracellular molecular state and the function of TLP through its property of binding to TLP.
Specifications
Specifications
GTF2A1
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
P52655
TF2A1, TFIIA, TFIIA alpha, p55, TFIIA-42, TFIIAL, general transcription factor IIA subunit 1, glucose regulated protein, 58kD pseudogene, transcription initiation factor IIA subunit 1, transcription initiation factor TFIIA 42 kDa subunit
Mouse
IgG1, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
2957
ELISA, Western Blot
2H5
Unconjugated
GTF2A1
Liquid
GTF2A1
GTF2A1 (NP_056943, 1 a.a. ∽ 63 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human