Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTF2A2, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127702

Catalog No. PIPA127702

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

TFIIA is a component of the transcription machinery of RNA polymerase II and plays an important role in transcriptional activation. Interacts with TBP (the TATA-binding protein).

PA1-27702 detects GTF2A2 from human and rat samples.PA1-27702 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Transcription initiation factor IIA gamma chain.
Specifications

Specifications

GTF2A2
Polyclonal
PBS with no preservative
P52657, O08950
HsT18745, TF2A2, TFIIA
Chicken
IgY
50μg
Store at -20°C
Primary
2958
Western Blot
Unconjugated
GTF2A2
Purified
GTF2A2
Synthetic peptide corresponding to human Transcription initiation factor IIA gamma chain.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit