Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

general transcription factor IIB (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human GTF2B protein.

Manufacturer:  Abnova Corporation H00002959B01

Catalog No. 89-005-483


Add to cart

Description

Description

This gene encodes the general transcription factor IIB, one of the ubiquitous factors required for transcription initiation by RNA polymerase II. The protein localizes to the nucleus where it forms a complex (the DAB complex) with transcription factors IID and IIA. Transcription factor IIB serves as a bridge between IID, the factor which initially recognizes the promoter sequence, and RNA polymerase II. [provided by RefSeq

Sequence: MASTSRLDALPRVTCPNHPDAILVEDYRAGDMICPECGLVVGDRVIDVGSEWRTFSNDKATKDPSRVGDSQNPLLSDGDLSTMIGKGTGAASFDEFGNSKYQNRRTMSSSDRAMMNAFKEITTMADRVNLPRNIVDRTNNLFKQVYEQKSLKGRANDAIASACLYIACRQEGVPRTFKEICAVSRISKKEIGRCFKLILKALETSVDLITTGDFMSRFCSNLCLPKQVQMAATHIARKAVELDLVPGRSPISVAAAAIYMASQASAEKRTQKEIGDIAGVADVTIRQSYRLIYPRAPDLFPTDFKFDTPVDKLPQL
Specifications

Specifications

general transcription factor IIB
Unconjugated
No additive
BC020597
TF2B|TFIIB
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
2959
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human GTF2B protein.
GTF2B
Serum
GTF2B
GTF2B (AAH20597, 1 a.a. ~ 316 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit