GTF2E2, Mouse, Clone: 2C3, Abnova™

Mouse monoclonal antibody raised against a full-length recombinant GTF2E2.

Manufacturer: Abnova Corporation H00002961M01

Catalog No. 89-022-417

  Check Availability
Add to Cart

    Specifications

    Format Liquid
    Formulation In 1X PBS, pH 7.4
    Gene GTF2E2
    Gene Accession No. BC030572
    Gene Alias FE, TF2E2, TFIIE-B
    Gene Symbols GTF2E2
    Applications Enzyme Linked Immunosorbent Assay,Immunofluorescence,Western Blotting
    Regulatory Status RUO
    Species Reactivity Human
    Storage Requirements Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
    Gene ID (Entrez) 2961
    Description general transcription factor IIE, polypeptide 2, beta 34kDa
    Antigen GTF2E2
    Host Species Mouse
    Immunogen GTF2E2 (AAH30572, 1 a.a. ∽ 291 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
    Isotype IgG1, κ
    Quantity 50μg
    Primary or Secondary Primary
    Monoclonal or Polyclonal Monoclonal
    Clone 2C3
    Conjugate Unlabeled
    Cross Reactivity Human

    Sequence: MDPSLLRERELFKKRALSTPVVEKRSASSESSSSSSKKKKTKVEHGGSSGSKQNSDHSNGSFNLKALSGSSGYKFGVLAKIVNYMKTRHQRGDTHPLTLDEILDETQHLDIGLKQKQWLMTEALVNNPKIEVIDGKYAFKPKYNVRDKKALLRLLDQHDQRGLGGILLEDIEEALPNSQKAVKALGDQILFVNRPDKKKILFFNDKSCQFSVDEEFQKLWRSVTVDSMDEEKIEEYLKRQGISSMQESGPKKVAPIQRRKKPASQKKRRFKTHNEHLAGVLKDYSDITSSK

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit