Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTP binding protein 1 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human GTPBP1 protein.

Manufacturer:  Abnova Corporation H00009567B01

Catalog No. 89-006-804


Add to cart

Description

Description

This gene is upregulated by interferon-gamma and encodes a protein that is a member of the AGP11/GTPBP1 family of GTP-binding proteins. A structurally similar protein has been found in mouse, where disruption of the gene for that protein had no observable phenotype. [provided by RefSeq

Sequence: MDEGCGETIYVIGQGSDGTEYGLSEADMEASYATVKSMAEQIEADVILLRERQEAGGRVRDYLVRKRVGDNDFLEVRVAVVGNVDAGKSTLLGVLTHGELDNGRGFARQKLFRHKHEIESGRTSSVGNDILGFDSEGNVVNKPDSHGGSLEWTKICEKSTKVITFIDLAGHEKYLKTTVFGMTGHLPDFCMLMVGSNAGIVGMTKEHLGLALALNVPVFVVVTKIDMCPANILQETLKLLQRLLKSPGCRKIPVLVQSKDDVIVTASNFSSERMCPIFQISNVTGENLDLLKMFLNLLSPRTSYREEEPAEFQIDDTYSVPGVGTVVSGTTLRGLIKLNDTLLLGPDPLGNFLSIAVKSIHRKRMPVKEVRGGQTASFALKKIKRSSIRKGMVMVSPRLNPQASWEFEAEILVLHHPTTISPRYQAMVHCGSIRQTATILSMDKDCLRTGDKATVHFRFIKTPEYLHIDQRLVFREGRTKAVGTITKLLQTTNNSPMNSKPQQIKMQSTKKGPLTKRDEGGPSGGPAVGAPPPGDEASSVGAGQPAASSNLQPQPKPSSGGRRRGGQRHKVKSQGACVTPASGC
Specifications

Specifications

GTP binding protein 1
Unconjugated
No additive
ENST00000335466
GP-1|GP1|HSPC018|MGC20069
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
9567
Western Blot
Mouse polyclonal antibody raised against a full-length human GTPBP1 protein.
GTPBP1
Serum
GTPBP1
GTPBP1 (ENSP00000334787, 1 a.a. ~ 584 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit