Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTPBP2 Rabbit anti-Hamster, Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523726

Catalog No. PIPA523726

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

GTP-binding proteins, or G proteins, constitute a superfamily capable of binding GTP or GDP. G proteins are activated by binding GTP and are inactivated by hydrolyzing GTP to GDP. This general mechanism enables G proteins to perform a wide range of biologic activities.
Specifications

Specifications

GTPBP2
Polyclonal
PBS with 0.09% sodium azide
Q9BX10, Q3UJK4
GTP-binding protein 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100758529, 54676, 56055
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
GTPBP2
Liquid
GTPBP2
KLH conjugated synthetic peptide between 536-564 amino acids from the C-terminal region of human GTPBP2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Hamster, Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit