Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTPBP8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524092

Catalog No. PIPA524092


Add to cart

Description

Description

This antibody is predicted to react with bovine based on sequence homology.

Specifications

Specifications

GTPBP8
Polyclonal
PBS with 0.09% sodium azide
Q8N3Z3
HSPC135, GTP-binding protein 8
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
29083
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
GTPBP8
Liquid
GTPBP8
KLH conjugated synthetic peptide between 122-150 amino acids from the central region of human GTPBP8
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit