Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-GTSE1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP130058

Catalog No. NBP130058


Add to cart

Description

Description

GTSE1 Polyclonal antibody specifically detects GTSE1 in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative).
Specifications

Specifications

GTSE1
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation
Affinity Purified
GTSE1
Maps to a region between residue 670 and 720 of human G-2 and S-phase expressed 1 using the numbering given in entry NP_057510.2 (GeneID 51512).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human
Immunoprecipitation, Western Blot
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
B99, G2 and S phase-expressed protein 1, G-2 and S-phase expressed 1, GTSE-1, Protein B99 homolog
Rabbit
IgG
0.1 ml
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary
51512
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only