Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GBP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA522238
Description
Recommended positive controls: A431, HeLa, HepG2, Molt-4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GBP3 Polyclonal antibody specifically detects GBP3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
GBP3 | |
Polyclonal | |
Unconjugated | |
GBP3 | |
GBP3; GBP-3; GTP-binding protein 3; guanine nucleotide-binding protein 3; guanylate binding protein 3; guanylate-binding protein 3; guanylate-binding protein 3 delta C; RP4-531M19.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2635 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.81 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9H0R5 | |
GBP3 | |
Recombinant fragment corresponding to a region within amino acids 61 and 317 of GBP3 (Uniprot ID#Q9H0R5) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction