Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GUCY1A2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513927

Catalog No. PIPA513927


Add to cart

Description

Description

This antibody is predicted to react with rat based on sequence homology.

Soluble guanylate cyclase (sGC), a heterodimeric protein consisting of an alpha and a beta subunit, catalyzes the conversion of GTP to the second messenger cGMP and functions as the main receptor for nitric oxide and nitrovasodilator drugs.
Specifications

Specifications

GUCY1A2
Polyclonal
Unconjugated
GUCY1A2
Liquid
GUCY1A2
KLH conjugated synthetic peptide between 105-136 amino acids from the N-terminal region of human Guanylyl Cyclase alpha 2 (GUCY1A2)
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
2 mg/mL
PBS with 0.09% sodium azide
P33402
Guanylate cyclase soluble, alpha-2 chain; GCS-alpha-2; GUCY1A2; GUC1A2, GUCSA2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
234889, 2977
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit