Learn More
anti-Guinea Pig IgG, HRP, Isotype Control, Novus Biologicals™
Guinea Pig Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196964
Catalog No. NBP196964
Add to cart
Description
Description
IgG antibody specifically detects IgG in Guinea Pig samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
IgG
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: None. with No Preservative
immunoglobulin heavy constant gamma 1 (G1m marker)
Guinea Pig
Ion exchange chromatography
RUO
Isotype Control
3500
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.
HRP
Lyophilized
IGHG1
IgG
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Primary
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Guinea Pig IgG and anti-Guinea Pig Serum.
Guinea Pig