Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-GUINPIGIGG(H+L) AB BIOTIN  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52600044
Affinity purified polyclonal antibody to Guniea Pig IgG, heavy and light chains (whole IgG) made in Goat and biotinylated. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-72