Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-GW182, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP128702
Description
GW182 Polyclonal antibody specifically detects GW182 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).Specifications
GW182 | |
1.0 mg/mL | |
Immunoprecipitation 10 ug/mg lysate, Western Blot (Negative) | |
Q8NDV7 | |
CAG repeat protein 26, CAGH26FLJ22043, DKFZp666E117, EMSY interactor protein, Glycine-tryptophan protein of 182 kDa, GW182 autoantigen, GW182GW1, KIAA1460EDIE, MGC75384, Protein GW1, TNRC6, trinucleotide repeat containing 6, trinucleotide repeat containing 6A, trinucleotide repeat-containing gene 6A protein | |
Rabbit | |
IgG | |
Immunogen affinity purified | |
RUO | |
Primary | |
27327 |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
Affinity Purified | |
TNRC6A | |
The immunogen recognized by this antibody maps to a region between residue 1225 and 1275 of human glycine-tryptophan protein of 182 kDa (GW182) using the numbering given in entry NP_055309.2 (GeneID 27327). | |
210 kDa | |
100 ul | |
Store at 4C. Do not freeze. | |
Polyclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only