Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-cadherin Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511487

Catalog No. PIPA511487

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with chicken and mouse based on sequence homology.

CDH13 is a member of the cadherin superfamily. This protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region but, unlike the typical cadherin superfamily member, lacks the highly conserved cytoplasmic region. This particular cadherin is a putative mediator of cell-cell interaction in the heart and may act as a negative regulator of neural cell growth.
Specifications

Specifications

H-cadherin
Polyclonal
PBS with 0.09% sodium azide
P55290
Cadherin-13; Truncated cadherin; T-cadherin; T-cad; Heart cadherin; H-cadherin; P105; CDH13; CDHH
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1012
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
CDH13
Liquid
CDH13
KLH conjugated synthetic peptide between 589-617 amino acids from the C-terminal region of human H Cadherin (CDH13)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit