Learn More
Bio X Cell InVivoMab anti-human CD47, Clone: B6H12, Size: 5 MG
Supplier:  Bio X Cell BE00191R005MG
Encompass_Preferred
InVivoMab anti-human CD47, Clone: B6H12, Size: 5 MG, Isotype: mouse IgG1, Applications: in vitro CD47 neutralization, iin vivo CD47 neutralization in human tumor xenograft models or humanized mice, Flow cytometry
Catalog No. 50-139-2755