Learn More
Bio X Cell InVivoMab anti-human CD71, Clone: OKT9, Size: 25 MG
Supplier:  Bio X Cell BE0023R025MG
Encompass_Preferred
InVivoMab anti-human CD71, Clone: OKT9, Size: 25 MG, Isotype: mouse IgG1, Applications: Flow cytometry, Immunohistochemistry (frozen)
Catalog No. 50-139-2776
Due to product restrictions, please to purchase or view availability for this product.