Learn More
Bio X Cell InVivoMab anti-human CTLA-4 (CD152), Clone: BN13, Size: 25 MG
SDP
Supplier:  Bio X Cell BE0190R025MG
Encompass_Preferred
InVivoMab anti-human CTLA-4 (CD152), Clone: BN13, Size: 25 MG, Isotype: mouse IgG2a, Applications: in vitro CTLA-4 neutralization, Flow cytometry
Catalog No. 50-139-3164