Learn More
IL-6 Rat anti-Human, Clone: MQ2-39C3, eBioscience
Rat Monoclonal Antibody
Manufacturer: Invitrogen 14706881
Description
Description: The MQ2-39C3 antibody reacts with human interleukin-6 (IL-6), a 21-28 kDa cytokine secreted by a variety of cell types. Applications Reported: MQ2-39C3 has been reported for use in detection of human IL-6 by ELISA and immunoblotting (WB). The purified MQ2-39C3 can be used for blocking of fluorochrome conjugated MQ2-39C3 in intracellular flow cytometry experiments to serve as specificity control. Applications Tested: Biotinylated MQ2-39C3 is routinely tested by sandwich ELISA for analysis of human IL-6 in combination with MQ2-13A5 and can be used at 1 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered.
Interleukin 6 (IL-6) is a multifunctional 26 kD protein originally discovered in the medium of RNA-stimulated fibroblastoid cells. IL-6 appears to be directly involved in the responses that occur after infection and cellular injury, and it may prove to be important as IL-1 and TNF-a in regulating the acute phase response. IL-6 is reported to be produced by fibroblasts, activated T cells, activated monocytes or macrophages and endothelial cells. IL-6 acts upon a variety of cells including myeloid progenitor cells, T cells, B cells, and hepatocytes. In addition, IL-6 appears to interact with IL-2 in the proliferation of T lymphocytes. IL-6 potentiates the proliferative effect of IL-3 on multipotential hematopoietic progenitors. IL-6 plays a critical role in B-cell differentiation to plasma cells and is a potent growth factor for plasmacytoma and myeloma. IL-6 is a very useful culture supplement for the generation of a high number of antibody-producing hybridomas. Primarily produced at sites of acute and chronic inflammation, IL-6 is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of IL-6 is implicated in a wide variety of inflammation-associated disease states including diabetes mellitus and systemic juvenile rheumatoid arthritis.Specifications
IL-6 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P05231 | |
Interleukin-6, BSF2, HGF, HSF, IFNB2, IL-6, B-cell stimulatory factor 2, BSF-2, Interferon beta-2, Hybridoma growth factor, CTL differentiation factor | |
Rat | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
ELISA | |
MQ2-39C3 | |
Unconjugated | |
IL6 | |
Liquid | |
IL6 | |
IgG2a, kappa | |
50 μg | |
4° C | |
Primary | |
3569 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.