Learn More
MilliporeSigma™ anti-H+,K+-ATPase, alpha-Subunit (1-17), Polyclonal
Rabbit Polyclonal Antibody
Supplier:  MilliporeSigma™ 11910250UL
Catalog No. 11-910-250UL
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Specifically detects H+,K+-ATPase,,alpha;-Subunit (1-17) Clone: in Canine, Human, Mouse samples, and it is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immuno Blotting
Specifications
Specifications
H+,K+-ATPase,,alpha;-Subunit (1-17)
Polyclonal
Rabbit
50 μL
Primary
Canine, Human, Mouse
Undiluted
ELISA, Immunoblot, Immunoblot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Unconjugated
a synthetic peptide (MGKAENYELYQVELGPGC) corresponding to amino acids 1-17 of porcine H+, K+-ATPase, alpha;-subunit
RUO
Recognizes theα subunit of H+,K+ ATPase.
≤ -70°C
IgG
Product Suggestions
Product Suggestions