Learn More
Bio X Cell InVivoMab anti-human PD-L1 (B7-H1), Clone: 29E.2A3, Size: 5 MG
Supplier:  Bio X Cell BE0285R005MG
Encompass_Preferred
InVivoMab anti-human PD-L1 (B7-H1), Clone: 29E.2A3, Size: 5 MG, Isotype: mouse IgG2b, Applications: in vitro PD-L1 blockade, Functional assays, Immunohistochemistry (frozen), Flow cytometry
Catalog No. 50-139-3429