Learn More
Bio X Cell InVivoMab anti-human/rat/fish AChR, Clone: Mab35 (TIB-175), Size: 25 MG
SDP
Supplier:  Bio X Cell BE0123R025MG
Encompass_Preferred
InVivoMab anti-human/rat/fish AChR, Clone: Mab35 (TIB-175), Size: 25 MG, Isotype: rat IgG1, Applications: EAMG induction in rats, Immunofluorescence
Catalog No. 50-139-2986