Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H6PD Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524960

Catalog No. PIPA524960


Add to cart

Description

Description

This antibody is predicted to react with bovine, mouse, porcine, rat and Xenopus based on sequence homology.

H6PD is 2 forms of glucose-6-phosphate dehydrogenase. G form is X-linked and H form, encoded by this gene, is autosomally linked. This H form shows activity with other hexose-6-phosphates, especially galactose-6-phosphate, whereas the G form is specific for glucose-6-phosphate. Both forms are present in most tissues, but H form is not found in red cells.
Specifications

Specifications

H6PD
Polyclonal
PBS with 0.09% sodium azide
O95479, Q8CFX1
G6PDH, GDH, 6-phosphogluconolactonase, G6PD, H form, GDH/6PGL endoplasmic bifunctional protein, glucose 1- dehydrogenase, glucose dehydrogenase, glucose dehyrogenase, glucose-6-phosphate dehydrogenase, salivary
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100198, 9563
Immunohistochemistry, Western Blot
Unconjugated
H6PD
Liquid
H6PD
KLH conjugated synthetic peptide between 743-769 amino acids from the C-terminal region of human H6PD
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit