Learn More
Bio X Cell ReadyTag anti-HA, Clone: 12CA5, Size: 25 MG
SDP
Supplier:  Bio X Cell RT0268R025MG
Encompass_Preferred
ReadyTag anti-HA, Clone: 12CA5, Size: 25 MG, Isotype: mouse IgG2b, Applications: ELISA, Western blot, Immunoprecipitation, Immunofluorescence
Catalog No. 50-139-3665