Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA Tag Rabbit anti-Tag, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1985

Catalog No. PA1985


Add to cart

Description

Description

PA1-985 detects HA tagged proteins. This antibody is specific for the HA tag and does not detect other proteins. PA1-985 has been successfully used in Western blot procedures. By Western blot, this antibody detects the presence of HA-tagged proteins. The PA1-985 immunogen is a synthetic peptide corresponding to the residues Y P Y D V P D Y A of the HA epitope of influenza hemagglutinin.

HA Tag (hemagglutinin tag) is an epitope tag that provides a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. HA (hemagglutinin) is a surface glycoprotein derived from influenza hemagglutinin, and is commonly expressed as a tag in conjunction with proteins to aid in identification and purification of the tagged protein for functional analysis. HA tag antibodies can be used to facilitate protein detection or co-immunopreciptation of tagged proteins.
Specifications

Specifications

HA Tag
Polyclonal
Unconjugated
Liquid
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Tag
Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
Human influenza hemagglutinin; hemagglutinin; HA-tag; HA; HA Epitope Tag
Synthetic peptide corresponding to the residues Y P Y D V P D Y A of the HA epitope of influenza hemagglutinin.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit