Learn More
UbpbioSupplier Diversity Partner ANTI-HA, MOUSE 100UG DFS Item
Manufacturer:  UbpbioSupplier Diversity Partner Y1071
Antibodies – Anti-HA (UBPBio catalog# Y1071) – Mouse monoclonal antibody (clone HA.C5, IgG3). Used for immunoblotting at 1:1,000-3,000 dilution for detection of 5 ng HA-tagged proteins. 1:500-2,000 dilution for immunostaining, 1-3 ug HA antibody/reaction for immunoprecipitation.
Catalog No. 50-112-6922