Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HAAO Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP132330
Description
HAAO Polyclonal specifically detects HAAO in Human, Mouse samples. It is validated for Western Blot.Specifications
HAAO | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
HAAO | |
Recombinant protein encompassing a sequence within the center region of human HAAO. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Tested in human. Predicted cross-reactivity based on sequence identity: Bovine (89%). Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
1.0mg/mL | |
Western Blot 1:500-1:10000 | |
3-HAO, 3-hydroxyanthranilate 3,4-dioxygenase, 3-hydroxyanthranilic acid dioxygenase, EC 1.13.11.6,3-hydroxyanthranilate oxygenase, HAD, HAO | |
Rabbit | |
33 kDa | |
100 μL | |
Lipid and Metabolism | |
23498 | |
Human, Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction