Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAGH, Polyclonal, Invitrogen™

Catalog No. PIPA127688
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA127688 50 μg
1 options

Catalog No. PIPA127688

Supplier: Thermo Scientific PA127688

Chicken Polyclonal Antibody

Thiolesterase that catalyzes the hydrolysis of S-D-lactoyl-glutathione to form glutathione and D-lactic acid. The enzyme encoded by this gene is classified as a thiolesterase and is responsible for the hydrolysis of S-lactoyl-glutathione to reduced glutat

PA1-27688 detects HAGH from human, mouse, and rat samples.PA1-27688 may be successfully used in ELISA and Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27935 immunogen is a synthetic peptide corresponding to human Hydroxyacylglutathione hydrolase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAGH
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene HAGH
Gene Accession No. Q16775, Q99KB8, O35952
Gene Alias GLO2, GLX2, GLXII, HAGH1
Gene Symbols HAGH
Host Species Chicken
Immunogen Synthetic peptide corresponding to human Hydroxyacylglutathione hydrolase.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3029
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.