Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAMP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526470

Catalog No. PIPA526470


Add to cart

Description

Description

HAMP is involved in the maintece of iron homeostasis, and it is necessary for the regulation of iron storage in macrophages, and for intestinal iron absorption. The preproprotein is post-translationally cleaved into mature peptides of 20, 22 and 25 amino acids, and these active peptides are rich in cysteines, which form intramolecular bonds that stabilize their beta-sheet structures. These peptides exhibit antimicrobial activity.
Specifications

Specifications

HAMP
Polyclonal
PBS with 0.09% sodium azide
P81172
HEPC, HFE2B, LEAP1, PLTR, hepcidin, liver-expressed antimicrobial peptide 1, putative liver tumor regressor
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57817
Western Blot
Unconjugated
HAMP
Liquid
HAMP
KLH conjugated synthetic peptide between 17-46 amino acids from the central region of human HAMP
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit