Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Hamster IgG, AP, Isotype Control, Novus Biologicals™

Golden Syrian Hamster Isotype Control

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197109

Catalog No. NBP197109


Add to cart

Description

Description

IgG antibody specifically detects IgG in Golden Syrian Hamster samples. It is validated for Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifications

Specifications

IgG
0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0. Stabilizer: None. with No Preservative
immunoglobulin heavy constant gamma 1 (G1m marker)
Golden Syrian Hamster
Ion exchange chromatography
RUO
Isotype Control
3500
Golden Syrian Hamster
AP
Conjugated
IGHG1
IgG
1 mg
Store at 4C. Do not freeze.
Primary
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Hamster IgG, anti-Hamster Serum and anti-Alkaline Phosphatase (calf intestine).
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only