Learn More
anti-Hamster IgG, Fluorescein, Isotype Control, Novus Biologicals™
Golden Syrian Hamster Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196872
Catalog No. NBP196872
Add to cart
Description
Description
IgG antibody specifically detects IgG in Golden Syrian Hamster samples. It is validated for Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifications
Specifications
IgG
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Sodium Azide
immunoglobulin heavy constant gamma 1 (G1m marker)
Golden Syrian Hamster
Ion exchange chromatography
RUO
Isotype Control
3500
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.
Fluorescein
Lyophilized
IGHG1
IgG
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Primary
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Fluorescein, anti-GS Hamster IgG and anti-GS Hamster Serum. Greatly diminished reactivity will occur against anti-Armenian Hamster IgG.
Golden Syrian Hamster