Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabbit anti-Syrian Hamster IgG (H+L) Cross-Adsorbed Secondary Antibody, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Supplier: Invitrogen A18897
Description
Syrian Hamster IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Syrian Hamster IgG (H+L) Cross-Adsorbed in Hamster samples. It is validated for ELISA.Specifications
Syrian Hamster IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Affinity chromatography | |
RUO | |
Hamster | |
Liquid |
ELISA | |
> 4.5 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
Gamma Immunoglobins Heavy and Light chains | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction