Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Hamster IgG1, κ Isotype Control, Clone: A19-3, BD

Manufacturer:  BD Biosciences 563612

Catalog No. BDB563612


Add to cart

Description

Description

Host Species: Hamster
Clone: A19-3
Isotype: IgG1, kappa
Species Reactivity: Hu, Ms
Immunogen: TNP-keyhole limpet hemocyanin

Isotype Control

Specifications

Specifications

Control, Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
TNP-keyhole limpet hemocyanin
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions, and unreacted Alexa Fluor™ 647 was removed.
RUO
Isotype Control
Monoclonal
A19-3
Alexa
Affinity Purified
Armenian Hamster IgG1, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype Control
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit