Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Hamster IgG1, lambda1 Isotype Control, Clone: G235-2356, BD

Manufacturer:  BD Biosciences 563751

Catalog No. BDB563751


Add to cart

Description

Description

Host Species: Hamster
Clone: G235-2356
Isotype: IgG1, lambda2
Species Reactivity: Ms
Immunogen: C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes

Isotype Control

Specifications

Specifications

Control, Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Trinitrophenol-KLH
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV786 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV786 were removed.
RUO
Isotype Control
Monoclonal
G235-2356
BV786
Affinity Purified
Armenian Hamster IgG1, λ1
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype Control
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit