Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HAPLN3, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP224476

 View more versions of this product

Catalog No. NBP224476


Add to cart

Description

Description

HAPLN3 Polyclonal specifically detects HAPLN3 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

HAPLN3
0.5 mg/mL
Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Affinity Purified
HAPLN3
A portion of amino acids 1-50 of human HAPLN3 was used as the immunogen.
Immunogen affinity purified
RUO
Primary
145864
Human, Primate
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide
EXLD1, extracellular link domain containing, 1, HsT19883, hyaluronan and proteoglycan link protein 3
Rabbit
IgG
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_b), chimpanzee and monkey, 88% homologous in cow, 82% homologous in mouse and rat and 76% homologous in zebrafish.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only