Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAUS4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525153

Catalog No. PIPA525153

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

HAUS4 encodes a subunit of the centrosome complex termed the human augmin complex. The encoded protein localizes to the spindle microtubules and may play a role in mitotic spindle assembly and maintece of centrosome integrity during cell division.
Specifications

Specifications

HAUS4
Polyclonal
PBS with 0.09% sodium azide
Q9H6D7
C14orf94, HAUS augmin-like complex subunit 4, homologous to Augmin subunits
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54930
Immunohistochemistry, Western Blot
Unconjugated
HAUS4
Liquid
HAUS4
KLH conjugated synthetic peptide between 288-316 amino acids from the C-terminal region of human HAUS4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit