Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAUS5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525154

Catalog No. PIPA525154

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The function of this protein has not been specifically defined.
Specifications

Specifications

HAUS5
Polyclonal
PBS with 0.09% sodium azide
O94927
KIAA0841, dgt5, HAUS augmin-like complex subunit 5
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23354
Flow Cytometry, Immunohistochemistry
Unconjugated
HAUS5
Liquid
HAUS5
KLH conjugated synthetic peptide between 139-167 amino acids from the N-terminal region of human HAUS5
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit