Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAX-1 Mouse, Unlabeled, Clone: 52, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610825

 View more versions of this product

Catalog No. BDB610825


Add to cart

Description

Description

Cell-cell interactions in the immune system, mediated by membrane-bound antigen receptors, trigger a cascade of signaling mechanisms collectively known as the T-cell response. Upon antigen binding, cellular tyrosine kinases are activated resulting in the phosphorylation of HS1 or Lck-binding protein 1 (LckBP1), which is involved in thymic negative selection. HS1 is one of the earliest intracellular proteins to be tyrosine phosphorylated in response to antigen-receptor cross-linking in T cells. HAX-1 (HS1-associated protein X-1) is a 35kDa protein found associated with HS1 by co-immunoprecipitation assays and the two-hybrid system. HAX-1 is ubiquitously expressed and localized in the mitochondria, endoplasmic reticulum, and the nuclear envelope. The HS1-HAX-1 complex may be critical for signal transduction pathways initiated by antigen presentation to T cells.

Immunofluorescence, Immunoprecipitation, Western Blotting

Specifications

Specifications

HAX-1
52
Unconjugated
Affinity Purified
Human HAX-1 aa. 10-148
150μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.