Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HAX-1, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128796

Catalog No. NBP128796


Add to cart

Description

Description

HAX-1 Polyclonal antibody specifically detects HAX-1 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications

Specifications

HAX-1
0.5 mg/mL
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:2000
NP_006109.2
FLJ17042, FLJ93803, HAX-1, HCLS1 (and PKD2) associated protein, HCLS1 associated protein X-1, HCLS1-associated protein X-1, HCLSBP1, HS1 binding protein, HS1-associating protein X-1, HS1-binding protein 1, HS1BP1FLJ18492, HSP1BP-1, SCN3
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognize both reported isoforms (NP_006109.2 and NP_001018238.1).
ELISA, Western Blot
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
HAX1
Peptide with sequence C-TRHEADSSPRGDPES corresponding to internal region according to NP_006109.2, NP_001018238.1.
Immunogen affinity purified
RUO
Primary
10456
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only