Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

HBEGF Mouse anti-Human, Clone: 406316, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523914

Catalog No. PIMA523914



Description

Description

In direct ELISAs and Western blots, this antibody does not cross-react with recombit human (rh) Amphiregulin, rhBTC, rhHRG-alpha, rhHRG-beta, rhEGF, rhTGF-alpha, recombit mouse (rm) Epigen, or rmEpiregulin. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 µg of the antibody by the LAL method.

Specifications

Specifications

HBEGF
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q99075
DTR, DTS, DTSF, HEGFL
Mouse
IgG2A
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
1839
Neutralization, Western Blot
406316
Unconjugated
HBEGF
Lyophilized
HBEGF
Sf 21-derived recombit human HB-EGF Asp63-Leu148
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit