Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYST2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511253

Catalog No. PIPA511253

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

HBO1/MYST2 is a histone acetyltransferase which specifically represses AR-mediated transcription. It may play a role in DNA replication.
Specifications

Specifications

MYST2
Polyclonal
PBS with 0.09% sodium azide
O95251
Histone acetyltransferase MYST2; EC 2.3.1.48; MYST protein 2; MOZ, YBF2/SAS3, SAS2 and TIP60 protein 2; Histone acetyltransferase binding to ORC1; MYST2; HBO1; HBOa
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11143
Immunohistochemistry, Western Blot
Unconjugated
KAT7
Liquid
KAT7
KLH conjugated synthetic peptide between 99-130 amino acids from the N-terminal region of human HBO1/MYST2
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit