Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOB2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526359

Catalog No. PIPA526359

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

HCCA2 binds to and stimulates the kinase activity of the related human serine-threonine kinases NDR1 and NDR2.
Specifications

Specifications

MOB2
Polyclonal
PBS with 0.09% sodium azide
Q70IA6, Q8VI63
HCCA2, MOB2 Mps One Binder homolog, Mps one binder kinase activator-like 2, mob2 homolog
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
101513, 81532
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
MOB2
Liquid
MOB2
KLH conjugated synthetic peptide between 5-32 amino acids from the N-terminal region of human HCCA2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit